Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63656772-63657041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB3
Entrez Gene ID
6317
Gene Name
serpin family B member 3
Gene Aliases
HsT1196, SCC, SCCA-1, SCCA-PD, SCCA1, SSCA1, T4-A
UniGene
Hs.227948
Gene Chromosome Location
Chr.18: 63655197-63661963 on Build GRCh38
Transcripts
NM_006919.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1711473 c.711A>T p.K237N Chr.18: 63656888-63656888 on Build GRCh38
3527047 c.768+1G>A p.? Chr.18: 63656830-63656830 on Build GRCh38
3144536 c.748G>A p.E250K Chr.18: 63656851-63656851 on Build GRCh38
6896724 c.721C>T p.L241L Chr.18: 63656878-63656878 on Build GRCh38
5833575 c.758delG p.G253fs*14 Chr.18: 63656841-63656841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140767490 Chr.18: 63656968-63656968 on Build GRCh38
rs61733411 Chr.18: 63656980-63656980 on Build GRCh38
rs939413176 Chr.18: 63656965-63656965 on Build GRCh38
rs151099886 Chr.18: 63656931-63656931 on Build GRCh38
rs748141618 Chr.18: 63656920-63656926 on Build GRCh38
rs75355927 Chr.18: 63656816-63656816 on Build GRCh38
rs758713585 Chr.18: 63656808-63656808 on Build GRCh38
rs141652634 Chr.18: 63656933-63656933 on Build GRCh38
rs543275784 Chr.18: 63656822-63656822 on Build GRCh38
rs3786333 Chr.18: 63656799-63656799 on Build GRCh38
rs753471125 Chr.18: 63656979-63656979 on Build GRCh38
rs769060548 Chr.18: 63656955-63656955 on Build GRCh38
rs765803075 Chr.18: 63656969-63656969 on Build GRCh38
rs750177130 Chr.18: 63656893-63656893 on Build GRCh38
rs746000475 Chr.18: 63656839-63656839 on Build GRCh38
rs778215018 Chr.18: 63656814-63656814 on Build GRCh38
rs757663889 Chr.18: 63656990-63656990 on Build GRCh38
rs143869105 Chr.18: 63656846-63656846 on Build GRCh38
rs200420977 Chr.18: 63656945-63656945 on Build GRCh38
rs749454876 Chr.18: 63657017-63657017 on Build GRCh38
rs758724951 Chr.18: 63656913-63656913 on Build GRCh38
rs199672164 Chr.18: 63657014-63657014 on Build GRCh38
rs778693875 Chr.18: 63656908-63656908 on Build GRCh38
rs774287307 Chr.18: 63656958-63656958 on Build GRCh38
rs781235774 Chr.18: 63656837-63656837 on Build GRCh38
rs749925376 Chr.18: 63656886-63656886 on Build GRCh38
rs762758799 Chr.18: 63656861-63656861 on Build GRCh38
rs377354078 Chr.18: 63657007-63657007 on Build GRCh38
rs61733410 Chr.18: 63656881-63656881 on Build GRCh38
rs780280279 Chr.18: 63656940-63656940 on Build GRCh38
rs750922283 Chr.18: 63657000-63657000 on Build GRCh38
rs766008369 Chr.18: 63656892-63656892 on Build GRCh38
rs146881818 Chr.18: 63656863-63656863 on Build GRCh38
rs139434361 Chr.18: 63656870-63656870 on Build GRCh38
rs144394860 Chr.18: 63656894-63656894 on Build GRCh38
rs200999787 Chr.18: 63656868-63656868 on Build GRCh38
rs924334593 Chr.18: 63656810-63656810 on Build GRCh38
rs560994708 Chr.18: 63656978-63656978 on Build GRCh38
rs762253199 Chr.18: 63656961-63656961 on Build GRCh38
rs148723757 Chr.18: 63656845-63656845 on Build GRCh38
rs767959004 Chr.18: 63656998-63656998 on Build GRCh38
rs773719131 Chr.18: 63656970-63656970 on Build GRCh38
rs760152632 Chr.18: 63656891-63656891 on Build GRCh38
rs554883652 Chr.18: 63656930-63656930 on Build GRCh38
rs765812385 Chr.18: 63656975-63656975 on Build GRCh38
rs778123166 Chr.18: 63656917-63656917 on Build GRCh38
rs751976916 Chr.18: 63656919-63656919 on Build GRCh38
rs770938183 Chr.18: 63656828-63656828 on Build GRCh38
rs117098817 Chr.18: 63656966-63656966 on Build GRCh38
rs772074402 Chr.18: 63656957-63656959 on Build GRCh38

View Full Product Details