The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006041.2 | NP_006032.1 | 2 - 2 | 2179 | 95 |
GenBank mRNA | BC063301.1 | - | 2 - 2 | 2096 | 95 |