Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUC2
Entrez Gene ID
4583
Gene Name
mucin 2, oligomeric mucus/gel-forming
Gene Aliases
MLP, MUC-2, SMUC
Chromosome Location
Chr.11: 1074875 - 1110508 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002457.3 NP_002448.3 - 1395 57
GenBank mRNA BQ340542.1 - 2 - 3 163 57

View Full Product Details