The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319674.1 | NP_001306603.1 | 5 - 6 | 584 | 98 |
NM_006582.3 | NP_006573.2 | 5 - 6 | 614 | 98 | |
NM_024482.2 | NP_077808.1 | 5 - 6 | 581 | 98 | |
XM_011540518.2 | XP_011538820.1 | 5 - 6 | 717 | 98 | |
XM_011540519.2 | XP_011538821.1 | 5 - 6 | 557 | 98 | |
XM_011540521.2 | XP_011538823.1 | 5 - 6 | 686 | 98 | |
XM_011540522.2 | XP_011538824.1 | 5 - 6 | 526 | 98 | |
XM_017000087.1 | XP_016855576.1 | 5 - 6 | 732 | 98 | |
GenBank mRNA | AF099013.1 | - | 4 - 5 | 478 | 98 |
AF173868.1 | - | 5 - 6 | 509 | 98 | |
AF203694.1 | - | 5 - 6 | 508 | 98 | |
AK000892.1 | - | 5 - 6 | 580 | 98 | |
AK023126.1 | - | 5 - 6 | 501 | 98 | |
AK315195.1 | - | 5 - 6 | 560 | 98 | |
AK315436.1 | - | 5 - 6 | 518 | 98 | |
BC001473.2 | - | 5 - 6 | 531 | 98 |