The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319674.1 | NP_001306603.1 | 7 - 8 | 870 | 77 |
NM_006582.3 | NP_006573.2 | 7 - 8 | 900 | 77 | |
NM_024482.2 | NP_077808.1 | 7 - 8 | 867 | 77 | |
XM_011540518.2 | XP_011538820.1 | 7 - 8 | 1003 | 77 | |
XM_011540519.2 | XP_011538821.1 | 7 - 8 | 843 | 77 | |
XM_011540521.2 | XP_011538823.1 | 7 - 8 | 972 | 77 | |
XM_011540522.2 | XP_011538824.1 | 7 - 8 | 819 | 77 | |
XM_017000087.1 | XP_016855576.1 | 7 - 8 | 1018 | 77 | |
GenBank mRNA | AF099013.1 | - | 6 - 7 | 764 | 77 |
AF173868.1 | - | 7 - 8 | 795 | 77 | |
AF203694.1 | - | 7 - 8 | 794 | 77 | |
AK000892.1 | - | 7 - 8 | 866 | 77 | |
AK023126.1 | - | 7 - 8 | 787 | 77 | |
AK315195.1 | - | 7 - 8 | 846 | 77 | |
AK315436.1 | - | 7 - 8 | 804 | 77 | |
BC001473.2 | - | 7 - 8 | 817 | 77 |