Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFSD11
Entrez Gene ID
79157
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
ET
Chromosome Location
Chr.17: 76736565 - 76803805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242532.1 NP_001229461.1 11 - 12 1903 87
NM_001242533.1 NP_001229462.1 12 - 13 1787 87
NM_001242534.1 NP_001229463.1 12 - 13 1649 87
NM_001242535.1 NP_001229464.1 12 - 13 1598 87
NM_001242536.1 NP_001229465.1 10 - 11 1631 87
NM_001242537.1 NP_001229466.1 10 - 11 1442 87
NM_024311.3 NP_077287.1 12 - 13 1505 87
XM_011525236.2 XP_011523538.1 11 - 12 1167 87
XM_011525237.2 XP_011523539.2 12 - 13 1610 87
XM_011525238.2 XP_011523540.2 12 - 13 1210 87
XM_011525239.2 XP_011523541.2 12 - 13 1180 87
XM_011525240.2 XP_011523542.2 12 - 13 1206 87
XM_011525241.2 XP_011523543.2 12 - 13 1216 87
XM_011525242.1 XP_011523544.1 11 - 12 1167 87
XM_011525244.1 XP_011523546.1 11 - 12 1167 87
XM_011525247.2 XP_011523549.2 12 - 13 1650 87
XM_017025065.1 XP_016880554.1 12 - 13 1238 87
XM_017025066.1 XP_016880555.1 12 - 13 1180 87
XM_017025067.1 XP_016880556.1 12 - 13 1238 87
XM_017025068.1 XP_016880557.1 12 - 13 1650 87
XM_017025069.1 XP_016880558.1 10 - 11 1338 87
XM_017025070.1 XP_016880559.1 10 - 11 1338 87
XM_017025071.1 XP_016880560.1 4 - 5 527 87
GenBank mRNA AF015184.1 - 12 - 13 1217 87
AF015185.1 - 10 - 11 1248 87
AF015186.1 - 12 - 13 1361 87
AK025849.1 - 11 - 12 1040 87
AK308058.1 - 10 - 11 1086 87
BC002753.2 - 12 - 13 1305 87
BC062563.1 - 12 - 13 1398 87

View Full Product Details