The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242533.1 | NP_001229462.1 | 1 - 2 | 664 | 80 |
NM_001242536.1 | NP_001229465.1 | 1 - 2 | 664 | 80 | |
XM_011525237.2 | XP_011523539.2 | 1 - 2 | 487 | 80 | |
XM_011525247.2 | XP_011523549.2 | 1 - 2 | 487 | 80 | |
XM_017025068.1 | XP_016880557.1 | 1 - 2 | 487 | 80 | |
GenBank mRNA | AF015184.1 | - | 1 - 2 | 94 | 80 |
AK308058.1 | - | 1 - 2 | 119 | 80 |