Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM21C
Entrez Gene ID
253725
Gene Name
family with sequence similarity 21 member C
Gene Aliases
FAM21A, VPEF
Chromosome Location
Chr.10: 45727170 - 45792964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011539568.1 XP_011537870.1 16 - 17 1626 111
XM_017016016.1 XP_016871505.1 16 - 17 1626 111
XM_017016017.1 XP_016871506.1 15 - 16 1554 111
XM_017016025.1 XP_016871514.1 7 - 8 696 111
GenBank mRNA AK293929.1 - 12 - 13 1197 111

Gene Symbol
FAM21A
Entrez Gene ID
387680
Gene Name
family with sequence similarity 21 member A
Gene Aliases
FAM21B, bA56A21.1, bA98I6.1
Chromosome Location
Chr.10: 50067888 - 50133509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005269807.3 XP_005269864.2 16 - 17 1607 111
XM_017016229.1 XP_016871718.1 16 - 17 1607 111
XM_017016232.1 XP_016871721.1 15 - 16 1535 111
XM_017016234.1 XP_016871723.1 16 - 17 1629 111

View Full Product Details