The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242536.1 | NP_001229465.1 | 5 - 6 | 1072 | 86 |
NM_001242537.1 | NP_001229466.1 | 5 - 6 | 883 | 86 | |
GenBank mRNA | AF015185.1 | - | 5 - 6 | 689 | 86 |
AK308058.1 | - | 5 - 6 | 527 | 86 |