Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
Chromosome Location
Chr.15: 42723557 - 42737219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138477.2 NP_612486.2 21 - 22 2984 69
XM_005254176.4 XP_005254233.1 21 - 22 2899 69
XM_011521270.2 XP_011519572.1 21 - 22 2924 69
XM_011521271.2 XP_011519573.1 21 - 22 2921 69
XM_011521274.2 XP_011519576.1 18 - 19 1970 69
GenBank mRNA AF525398.1 - 21 - 22 2981 69
BC052568.1 - 21 - 22 2872 69
BC066640.1 - 21 - 22 2872 69

View Full Product Details