The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252338.2 | NP_001239267.1 | 10 - 11 | 1676 | 97 |
NM_001252339.2 | NP_001239268.1 | 10 - 11 | 1833 | 97 | |
NM_001794.4 | NP_001785.2 | 11 - 12 | 1861 | 97 | |
GenBank mRNA | AK091496.1 | - | 11 - 12 | 2057 | 97 |
AK124724.1 | - | 10 - 11 | 1816 | 97 | |
AK308345.1 | - | 10 - 11 | 1675 | 97 | |
AK315192.1 | - | 11 - 12 | 1815 | 97 | |
BC101651.1 | - | 11 - 12 | 1861 | 97 | |
BC112150.1 | - | 11 - 12 | 1861 | 97 | |
L34059.1 | - | 11 - 12 | 1787 | 97 |