Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
Chromosome Location
Chr.2: 112773915 - 112785398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000575.4 NP_000566.3 5 - 6 1458 128
GenBank mRNA AK314850.1 - 5 - 6 553 128
BC013142.2 - 4 - 5 503 128
BT007014.1 - 4 - 5 494 128
CR457414.1 - 4 - 5 494 128
M15329.1 - 4 - 5 499 128
X02531.1 - 4 - 5 494 128
X02851.1 - 5 - 6 552 128
X56086.1 - 5 - 6 520 128

View Full Product Details