Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR4
Entrez Gene ID
10785
Gene Name
WD repeat domain 4
Gene Aliases
TRM82, TRMT82
Chromosome Location
Chr.21: 42843075 - 42879865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260474.1 NP_001247403.1 1 - 2 176 65
NM_001260475.1 NP_001247404.1 1 - 2 176 65
NM_018669.5 NP_061139.2 1 - 2 176 65
NM_033661.4 NP_387510.1 1 - 2 176 65
XM_017028263.1 XP_016883752.1 1 - 2 103 65
XM_017028264.1 XP_016883753.1 1 - 2 103 65
GenBank mRNA AJ243912.1 - 1 - 2 120 65
AJ243913.1 - 1 - 2 120 65
AK292923.1 - 1 - 2 117 65
AK315008.1 - 1 - 2 119 65
BC006341.2 - 1 - 2 124 65
BG722541.1 - 1 - 2 150 65
BU541429.1 - 1 - 2 102 65
DC303088.1 - 1 - 2 176 65

View Full Product Details