The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260474.1 | NP_001247403.1 | 5 - 6 | 657 | 69 |
NM_001260475.1 | NP_001247404.1 | 5 - 6 | 605 | 69 | |
NM_001260476.1 | NP_001247405.1 | 5 - 6 | 505 | 69 | |
NM_001260477.1 | NP_001247406.1 | 5 - 6 | 712 | 69 | |
NM_018669.5 | NP_061139.2 | 5 - 6 | 657 | 69 | |
NM_033661.4 | NP_387510.1 | 5 - 6 | 657 | 69 | |
NR_048535.1 | 5 - 6 | 549 | 69 | ||
XM_011529433.1 | XP_011527735.1 | 5 - 6 | 920 | 69 | |
GenBank mRNA | AJ243912.1 | - | 6 - 7 | 601 | 69 |
AJ243913.1 | - | 6 - 7 | 601 | 69 | |
AK056343.1 | - | 5 - 6 | 712 | 69 | |
AK292923.1 | - | 5 - 6 | 598 | 69 | |
AK298015.1 | - | 5 - 6 | 549 | 69 | |
AK315008.1 | - | 5 - 6 | 548 | 69 | |
BC001074.2 | - | 5 - 6 | 484 | 69 | |
BC006341.2 | - | 5 - 6 | 605 | 69 | |
BU541429.1 | - | 5 - 6 | 583 | 69 |