The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176877.2 | NP_795352.2 | 28 - 29 | 3984 | 86 |
XM_005270341.3 | XP_005270398.1 | 28 - 29 | 3982 | 86 | |
XM_011540462.2 | XP_011538764.1 | 28 - 29 | 3982 | 86 | |
XM_011540463.2 | XP_011538765.1 | 28 - 29 | 4170 | 86 | |
XM_011540464.2 | XP_011538766.1 | 28 - 29 | 3982 | 86 | |
XM_011540465.2 | XP_011538767.1 | 28 - 29 | 3982 | 86 | |
XM_011540466.2 | XP_011538768.1 | 28 - 29 | 3982 | 86 | |
XM_011540467.2 | XP_011538769.1 | 28 - 29 | 3982 | 86 | |
XM_011540468.2 | XP_011538770.1 | 28 - 29 | 3982 | 86 | |
XM_016999998.1 | XP_016855487.1 | 28 - 29 | 3982 | 86 | |
XM_016999999.1 | XP_016855488.1 | 28 - 29 | 3982 | 86 | |
XM_017000000.1 | XP_016855489.1 | 23 - 24 | 3228 | 86 | |
XM_017000001.1 | XP_016855490.1 | 9 - 10 | 1059 | 86 | |
GenBank mRNA | AB044807.1 | - | 15 - 16 | 2263 | 86 |
AF397170.1 | - | 27 - 28 | 3870 | 86 | |
AJ001306.1 | - | 28 - 29 | 3984 | 86 | |
AJ224747.1 | - | 28 - 29 | 3984 | 86 | |
AJ224748.1 | - | 28 - 29 | 3984 | 86 | |
AK130492.1 | - | 8 - 9 | 946 | 86 | |
AK293519.1 | - | 2 - 3 | 300 | 86 | |
AK304355.1 | - | 23 - 24 | 3341 | 86 | |
AK308665.1 | - | 2 - 3 | 547 | 86 | |
BC144135.1 | - | 28 - 29 | 3936 | 86 |