The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176877.2 | NP_795352.2 | 3 - 4 | 301 | 67 |
XM_005270341.3 | XP_005270398.1 | 3 - 4 | 299 | 67 | |
XM_005270347.1 | XP_005270404.1 | 3 - 4 | 299 | 67 | |
XM_006710278.3 | XP_006710341.1 | 3 - 4 | 299 | 67 | |
XM_011540462.2 | XP_011538764.1 | 3 - 4 | 299 | 67 | |
XM_011540463.2 | XP_011538765.1 | 3 - 4 | 487 | 67 | |
XM_011540464.2 | XP_011538766.1 | 3 - 4 | 299 | 67 | |
XM_011540465.2 | XP_011538767.1 | 3 - 4 | 299 | 67 | |
XM_011540466.2 | XP_011538768.1 | 3 - 4 | 299 | 67 | |
XM_011540467.2 | XP_011538769.1 | 3 - 4 | 299 | 67 | |
XM_011540468.2 | XP_011538770.1 | 3 - 4 | 299 | 67 | |
XM_011540469.2 | XP_011538771.1 | 3 - 4 | 299 | 67 | |
XM_011540470.1 | XP_011538772.1 | 3 - 4 | 299 | 67 | |
XM_016999998.1 | XP_016855487.1 | 3 - 4 | 299 | 67 | |
XM_016999999.1 | XP_016855488.1 | 3 - 4 | 299 | 67 | |
GenBank mRNA | AF397170.1 | - | 2 - 3 | 187 | 67 |
AJ001306.1 | - | 3 - 4 | 301 | 67 | |
AJ224747.1 | - | 3 - 4 | 301 | 67 | |
AJ224748.1 | - | 3 - 4 | 301 | 67 | |
AK307713.1 | - | 3 - 4 | 298 | 67 | |
AK308428.1 | - | 3 - 4 | 298 | 67 | |
BC142661.1 | - | 3 - 4 | 284 | 67 | |
BC142723.1 | - | 3 - 4 | 284 | 67 | |
BC144135.1 | - | 2 - 3 | 233 | 67 |