Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAMP3
Entrez Gene ID
10067
Gene Name
secretory carrier membrane protein 3
Gene Aliases
C1orf3
Chromosome Location
Chr.1: 155255979 - 155262703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006711105.3 XP_006711168.1 4 - 5 941 122

View Full Product Details