Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASNS
Entrez Gene ID
440
Gene Name
asparagine synthetase (glutamine-hydrolyzing)
Gene Aliases
ASNSD, TS11
Chromosome Location
Chr.7: 97852117 - 97872542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178075.1 NP_001171546.1 9 - 10 1320 72
NM_001178076.1 NP_001171547.1 7 - 8 1086 72
NM_001178077.1 NP_001171548.1 7 - 8 1019 72
NM_001673.4 NP_001664.3 9 - 10 1394 72
NM_133436.3 NP_597680.2 9 - 10 1649 72
NM_183356.3 NP_899199.2 10 - 11 1672 72
GenBank mRNA AK000379.1 - 9 - 10 1617 72
AK302189.1 - 9 - 10 1214 72
AK302242.1 - 7 - 8 967 72
AK316224.1 - 7 - 8 1021 72
BC008723.2 - 10 - 11 1499 72
BC014621.2 - 9 - 10 1270 72
BT007113.1 - 7 - 8 1143 72
M15798.1 - 8 - 9 1190 72
M27396.1 - 9 - 10 1321 72

View Full Product Details