Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFIC
Entrez Gene ID
4782
Gene Name
nuclear factor I C
Gene Aliases
CTF, CTF5, NF-I, NFI
Chromosome Location
Chr.19: 3359563 - 3469217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001245002.1 NP_001231931.1 4 - 5 782 63
NM_001245004.1 NP_001231933.1 4 - 5 782 63
NM_001245005.1 NP_001231934.1 4 - 5 805 63
NM_005597.3 NP_005588.2 4 - 5 782 63
NM_205843.2 NP_995315.1 4 - 5 805 63
XM_005259564.3 XP_005259621.1 4 - 5 790 63
XM_006722759.2 XP_006722822.2 4 - 5 718 63
XM_017026834.1 XP_016882323.1 4 - 5 790 63
XM_017026835.1 XP_016882324.1 4 - 5 790 63
XM_017026836.1 XP_016882325.1 4 - 5 718 63
GenBank mRNA AK289885.1 - 4 - 5 718 63
AK297825.1 - 4 - 5 764 63
AK297867.1 - 4 - 5 766 63
AK304816.1 - 4 - 5 721 63
BC012120.1 - 4 - 5 774 63
CR536507.1 - 3 - 4 685 63
X12492.1 - 4 - 5 752 63
X92857.1 - 4 - 5 736 63

View Full Product Details