Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAS2R8
Entrez Gene ID
50836
Gene Name
taste 2 receptor member 8
Gene Aliases
T2R8, TRB5
Chromosome Location
Chr.12: 10806051 - 10806980 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023918.1 NP_076407.1 1 - 1 655 150
GenBank mRNA BC069084.1 - 1 - 1 655 150
BC069551.1 - 1 - 1 655 150
BC096701.1 - 1 - 1 655 150
BC096735.1 - 1 - 1 655 150
BC098167.1 - 1 - 1 655 150
BC098353.1 - 1 - 1 655 150

View Full Product Details