The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 34 - 35 | 3842 | 106 |
XM_011512110.2 | XP_011510412.1 | 34 - 35 | 3859 | 106 | |
XM_011512111.2 | XP_011510413.1 | 34 - 35 | 3859 | 106 | |
XM_011512113.2 | XP_011510415.1 | 34 - 35 | 3853 | 106 | |
XM_011512114.2 | XP_011510416.1 | 34 - 35 | 3859 | 106 | |
XM_011512115.2 | XP_011510417.1 | 33 - 34 | 3790 | 106 | |
XM_011512116.2 | XP_011510418.1 | 34 - 35 | 3772 | 106 | |
XM_011512118.2 | XP_011510420.1 | 33 - 34 | 3739 | 106 | |
XM_011512119.2 | XP_011510421.1 | 34 - 35 | 3859 | 106 | |
XM_011512120.2 | XP_011510422.1 | 33 - 34 | 3750 | 106 | |
XM_011512122.2 | XP_011510424.1 | 34 - 35 | 3860 | 106 | |
XM_011512124.2 | XP_011510426.1 | 18 - 19 | 3443 | 106 | |
XM_011512125.2 | XP_011510427.1 | 34 - 35 | 3860 | 106 | |
XM_011512126.2 | XP_011510428.1 | 34 - 35 | 3859 | 106 | |
XM_017005222.1 | XP_016860711.1 | 34 - 35 | 3856 | 106 | |
XM_017005223.1 | XP_016860712.1 | 17 - 18 | 2507 | 106 | |
XM_017005224.1 | XP_016860713.1 | 8 - 9 | 1299 | 106 |