The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293083.1 | NP_001280012.1 | 50 - 51 | 5792 | 60 |
XM_011512110.2 | XP_011510412.1 | 50 - 51 | 5836 | 60 | |
XM_011512111.2 | XP_011510413.1 | 50 - 51 | 5833 | 60 | |
XM_011512113.2 | XP_011510415.1 | 50 - 51 | 5830 | 60 | |
XM_011512114.2 | XP_011510416.1 | 50 - 51 | 5806 | 60 | |
XM_011512115.2 | XP_011510417.1 | 49 - 50 | 5767 | 60 | |
XM_011512116.2 | XP_011510418.1 | 50 - 51 | 5749 | 60 | |
XM_011512117.2 | XP_011510419.1 | 49 - 50 | 5731 | 60 | |
XM_011512118.2 | XP_011510420.1 | 49 - 50 | 5716 | 60 | |
XM_011512119.2 | XP_011510421.1 | 49 - 50 | 5674 | 60 | |
XM_011512120.2 | XP_011510422.1 | 49 - 50 | 5727 | 60 | |
XM_011512122.2 | XP_011510424.1 | 50 - 51 | 5837 | 60 | |
XM_011512124.2 | XP_011510426.1 | 34 - 35 | 5420 | 60 | |
XM_017005222.1 | XP_016860711.1 | 50 - 51 | 5833 | 60 | |
XM_017005223.1 | XP_016860712.1 | 33 - 34 | 4484 | 60 | |
XM_017005224.1 | XP_016860713.1 | 24 - 25 | 3276 | 60 | |
GenBank mRNA | AL133022.1 | - | 32 - 33 | 5195 | 60 |
AY461813.1 | - | 49 - 50 | 5818 | 60 | |
BC117324.1 | - | 16 - 17 | 2000 | 60 | |
BC126368.1 | - | 16 - 17 | 2003 | 60 | |
BC143825.1 | - | 16 - 17 | 2000 | 60 |