Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM2
Entrez Gene ID
23670
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 71683366 - 71769532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135820.1 NP_001129292.1 15 - 16 3121 104
NM_013390.2 NP_037522.1 16 - 17 3310 104
XM_005251869.4 XP_005251926.1 16 - 17 2943 104
GenBank mRNA AB037833.2 - 16 - 17 2903 104
AF137030.1 - 16 - 17 2918 104
AK075370.1 - 9 - 10 1038 104
BC140773.1 - 15 - 16 2623 104
BC144510.1 - 16 - 17 2778 104
BC146780.1 - 16 - 17 2903 104
BC171782.1 - 15 - 16 2624 104

View Full Product Details