The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014227.2 | NP_055042.1 | 4 - 5 | 369 | 66 |
XM_006724308.3 | XP_006724371.1 | 5 - 6 | 620 | 66 | |
XM_011530342.2 | XP_011528644.1 | 6 - 7 | 738 | 66 | |
XM_011530343.2 | XP_011528645.1 | 4 - 5 | 284 | 66 | |
XM_011530344.2 | XP_011528646.1 | 3 - 4 | 249 | 66 | |
XM_017028920.1 | XP_016884409.1 | 6 - 7 | 699 | 66 | |
GenBank mRNA | AJ133127.1 | - | 4 - 5 | 383 | 66 |
U41897.1 | - | 2 - 3 | 152 | 66 |