The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014227.2 | NP_055042.1 | 6 - 7 | 588 | 71 |
XM_006724308.3 | XP_006724371.1 | 7 - 8 | 839 | 71 | |
XM_011530342.2 | XP_011528644.1 | 8 - 9 | 957 | 71 | |
XM_011530343.2 | XP_011528645.1 | 6 - 7 | 503 | 71 | |
XM_011530344.2 | XP_011528646.1 | 5 - 6 | 468 | 71 | |
XM_017028920.1 | XP_016884409.1 | 8 - 9 | 918 | 71 | |
GenBank mRNA | AJ133127.1 | - | 6 - 7 | 602 | 71 |
U41897.1 | - | 4 - 5 | 371 | 71 |