The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130847.3 | NP_001124319.1 | 7 - 8 | 1013 | 106 |
NM_004208.3 | NP_004199.1 | 7 - 8 | 1013 | 106 | |
NM_145812.2 | NP_665811.1 | 7 - 8 | 1001 | 106 | |
NR_132647.1 | 6 - 7 | 870 | 106 | ||
GenBank mRNA | AF100928.1 | - | 7 - 8 | 824 | 106 |
AF131759.1 | - | 4 - 5 | 365 | 106 | |
AK000775.1 | - | 7 - 8 | 849 | 106 | |
AK314446.1 | - | 7 - 8 | 801 | 106 | |
AL049703.1 | - | 7 - 8 | 866 | 106 | |
AY795556.1 | - | 5 - 6 | 453 | 106 | |
BC111065.1 | - | 7 - 8 | 944 | 106 | |
BC139738.1 | - | 7 - 8 | 917 | 106 | |
CR457379.1 | - | 7 - 8 | 782 | 106 | |
DQ016498.1 | - | 7 - 8 | 790 | 106 | |
DQ016500.1 | - | 6 - 7 | 647 | 106 |