The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032287.2 | NP_001027458.1 | 1 - 2 | 250 | 108 |
NM_001127362.1 | NP_001120834.1 | 1 - 2 | 250 | 108 | |
NM_003297.3 | NP_003288.2 | 1 - 2 | 250 | 108 | |
XM_005269133.2 | XP_005269190.1 | 1 - 2 | 237 | 108 | |
XM_006719584.2 | XP_006719647.1 | 1 - 2 | 237 | 108 | |
XM_011538716.2 | XP_011537018.1 | 1 - 2 | 237 | 108 | |
XM_011538717.2 | XP_011537019.1 | 1 - 2 | 237 | 108 | |
GenBank mRNA | AK291319.1 | - | 1 - 2 | 214 | 108 |
BC026074.1 | - | 1 - 2 | 196 | 108 | |
BC040141.1 | - | 1 - 2 | 202 | 108 | |
DB266298.1 | - | 1 - 2 | 250 | 108 |