Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016260.2 NP_057344.2 4 - 5 577 78
XM_005246384.4 XP_005246441.1 4 - 5 687 78
XM_005246385.3 XP_005246442.1 4 - 5 717 78
XM_011510802.2 XP_011509104.1 5 - 6 759 78
XM_011510803.2 XP_011509105.1 5 - 6 608 78
XM_011510804.2 XP_011509106.1 5 - 6 531 78
XM_011510805.2 XP_011509107.1 5 - 6 620 78
XM_011510807.2 XP_011509109.1 5 - 6 795 78
XM_011510808.2 XP_011509110.1 5 - 6 645 78
XM_011510809.2 XP_011509111.1 5 - 6 657 78
XM_011510810.2 XP_011509112.1 5 - 6 571 78
XM_011510811.2 XP_011509113.1 4 - 5 692 78
XM_011510812.2 XP_011509114.1 4 - 5 553 78
XM_011510814.2 XP_011509116.1 4 - 5 1051 78
XM_011510816.2 XP_011509118.1 2 - 3 500 78
XM_011510817.2 XP_011509119.1 4 - 5 579 78
XM_011510818.2 XP_011509120.1 2 - 3 6154 78
XM_017003591.1 XP_016859080.1 2 - 3 1042 78
GenBank mRNA AF130863.1 - 4 - 5 577 78
AK301503.1 - 4 - 5 573 78
AY587062.1 - 5 - 6 561 78
AY587063.1 - 5 - 6 523 78
AY587064.1 - 3 - 4 408 78
AY587068.1 - 3 - 4 408 78

View Full Product Details