Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079526.1 NP_001072994.1 8 - 9 1096 62
NM_016260.2 NP_057344.2 7 - 8 1027 62
XM_005246384.4 XP_005246441.1 7 - 8 1137 62
XM_005246385.3 XP_005246442.1 7 - 8 1167 62
XM_005246386.4 XP_005246443.1 7 - 8 1018 62
XM_011510802.2 XP_011509104.1 8 - 9 1209 62
XM_011510803.2 XP_011509105.1 8 - 9 1058 62
XM_011510804.2 XP_011509106.1 8 - 9 981 62
XM_011510805.2 XP_011509107.1 8 - 9 1070 62
XM_011510807.2 XP_011509109.1 8 - 9 1245 62
XM_011510808.2 XP_011509110.1 8 - 9 1095 62
XM_011510809.2 XP_011509111.1 8 - 9 1107 62
XM_011510810.2 XP_011509112.1 8 - 9 1021 62
XM_011510811.2 XP_011509113.1 7 - 8 1142 62
XM_011510812.2 XP_011509114.1 7 - 8 1003 62
XM_011510814.2 XP_011509116.1 7 - 8 1501 62
XM_011510815.2 XP_011509117.1 7 - 8 949 62
XM_011510816.2 XP_011509118.1 5 - 6 950 62
XM_011510818.2 XP_011509120.1 5 - 6 6604 62
XM_017003589.1 XP_016859078.1 7 - 8 1089 62
XM_017003590.1 XP_016859079.1 7 - 8 1059 62
XM_017003591.1 XP_016859080.1 5 - 6 1492 62
GenBank mRNA AF130863.1 - 7 - 8 1027 62
AK301503.1 - 7 - 8 1023 62
AY008294.1 - 1 - 2 1491 62
AY587062.1 - 8 - 9 1011 62
AY587063.1 - 8 - 9 973 62
AY587064.1 - 8 - 9 1057 62
AY587065.1 - 7 - 8 864 62
AY587067.1 - 6 - 7 675 62
AY587068.1 - 7 - 8 935 62
AY587069.1 - 6 - 7 780 62
AY587070.1 - 4 - 5 423 62
AY587071.1 - 5 - 6 642 62
AY587073.1 - 7 - 8 1037 62
BC028936.1 - 8 - 9 1096 62

View Full Product Details