Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUNDC3B
Entrez Gene ID
154661
Gene Name
RUN domain containing 3B
Gene Aliases
RPIB9, RPIP9
Chromosome Location
Chr.7: 87627656 - 87832298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005250156.2 XP_005250213.1 9 - 10 1505 77
XM_011515826.1 XP_011514128.1 10 - 11 1556 77
XM_017011764.1 XP_016867253.1 7 - 8 980 77
GenBank mRNA AK293607.1 - 9 - 10 1321 77

View Full Product Details