The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002111.8 | NP_002102.4 | 10 - 11 | 1479 | 67 |
GenBank mRNA | AB016794.1 | - | 10 - 11 | 1479 | 67 |
L12392.1 | - | 10 - 11 | 1649 | 67 |