Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTT
Entrez Gene ID
3064
Gene Name
huntingtin
Gene Aliases
HD, IT15
Chromosome Location
Chr.4: 3074510 - 3243960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002111.8 NP_002102.4 60 - 61 8418 73
GenBank mRNA AB016794.1 - 60 - 61 8418 73
AB209506.1 - 46 - 47 9395 73
AK290544.1 - 5 - 6 631 73
KJ535072.1 - 5 - 6 619 73
L12392.1 - 60 - 61 8588 73
L20431.1 - 5 - 6 588 73

View Full Product Details