Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DERL1
Entrez Gene ID
79139
Gene Name
derlin 1
Gene Aliases
DER-1, DER1
Chromosome Location
Chr.8: 123013164 - 123042423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK303826.1 - 1 - 2 329 62

View Full Product Details