Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF2
Entrez Gene ID
22807
Gene Name
IKAROS family zinc finger 2
Gene Aliases
ANF1A2, HELIOS, ZNF1A2, ZNFN1A2
Chromosome Location
Chr.2: 212999684 - 213152753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011510817.2 XP_011509119.1 6 - 7 876 89
XM_011510819.2 XP_011509121.1 6 - 7 865 89
XM_017003592.1 XP_016859081.1 6 - 7 797 89
GenBank mRNA AY587066.1 - 5 - 6 627 89

View Full Product Details