Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKG2
Entrez Gene ID
5593
Gene Name
protein kinase, cGMP-dependent, type II
Gene Aliases
PRKGR2, cGK2, cGKII
Chromosome Location
Chr.4: 81086874 - 81217837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282480.1 NP_001269409.1 10 - 11 1045 146
NM_001282481.1 NP_001269410.1 9 - 10 919 146
NM_001282482.1 NP_001269411.1 9 - 10 832 146
NM_001282483.1 NP_001269412.1 9 - 10 896 146
NM_001282485.1 NP_001269414.1 14 - 15 1883 146
NM_006259.2 NP_006250.1 14 - 15 1970 146
XM_005263126.3 XP_005263183.1 14 - 15 2405 146
XM_017008413.1 XP_016863902.1 14 - 15 2407 146
XM_017008414.1 XP_016863903.1 14 - 15 2402 146
XM_017008415.1 XP_016863904.1 14 - 15 1812 146
XM_017008416.1 XP_016863905.1 14 - 15 2324 146
XM_017008417.1 XP_016863906.1 13 - 14 1701 146
GenBank mRNA AK296979.1 - 9 - 10 906 146
AK297673.1 - 14 - 15 1830 146
AK316140.1 - 10 - 11 1045 146
D70899.1 - 13 - 14 1775 146
X94612.1 - 13 - 14 1789 146

View Full Product Details