Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SGSM3
Entrez Gene ID
27352
Gene Name
small G protein signaling modulator 3
Gene Aliases
MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
Chromosome Location
Chr.22: 40370530 - 40410289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301849.1 NP_001288778.1 15 - 16 1796 112
NM_015705.5 NP_056520.2 16 - 17 2019 112
XM_005261572.2 XP_005261629.1 16 - 17 2046 112
XM_005261573.3 XP_005261630.1 16 - 17 2051 112
XM_005261574.4 XP_005261631.1 16 - 17 2051 112
XM_005261575.1 XP_005261632.1 15 - 16 1914 112
XM_011530158.1 XP_011528460.1 15 - 16 1870 112
XM_011530159.1 XP_011528461.1 16 - 17 1934 112
XM_011530160.1 XP_011528462.1 15 - 16 1851 112
XM_011530161.1 XP_011528463.1 16 - 17 2043 112
XM_011530162.2 XP_011528464.1 16 - 17 1929 112
XM_017028775.1 XP_016884264.1 15 - 16 1838 112
XM_017028776.1 XP_016884265.1 15 - 16 1907 112
XM_017028777.1 XP_016884266.1 15 - 16 1907 112
XM_017028778.1 XP_016884267.1 14 - 15 1770 112
XM_017028779.1 XP_016884268.1 15 - 16 1960 112
XM_017028780.1 XP_016884269.1 15 - 16 1949 112
GenBank mRNA AB196956.1 - 14 - 15 1788 112
AB275763.1 - 15 - 16 1892 112
AB449880.1 - 14 - 15 1781 112
AK000006.1 - 6 - 7 2929 112
AK297599.1 - 15 - 16 1867 112
AK301042.1 - 15 - 16 1709 112
AY308849.1 - 16 - 17 1954 112
BC008078.2 - 16 - 17 1945 112
CR456441.1 - 15 - 16 1817 112

View Full Product Details