Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SGSM3
- Entrez Gene ID
-
27352
- Gene Name
- small G protein signaling modulator 3
- Gene Aliases
-
MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
- Chromosome Location
-
Chr.22: 40370530 - 40410289 on Build GRCh38
View Full Product Details