Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAMP3
Entrez Gene ID
9341
Gene Name
vesicle associated membrane protein 3
Gene Aliases
CEB
Chromosome Location
Chr.1: 7771269 - 7781432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004781.3 NP_004772.1 1 - 2 115 93
XM_017002841.1 XP_016858330.1 1 - 2 109 93
GenBank mRNA AK316558.1 - 1 - 2 107 93
BC005941.1 - 1 - 2 115 93

View Full Product Details