The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033723.1 | 4 - 5 | 77 | ||
GenBank mRNA | AW161751.1 | - | 2 - 3 | 137 | 77 |
CB052107.1 | - | 4 - 5 | 77 |