Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOM
Entrez Gene ID
2040
Gene Name
stomatin
Gene Aliases
BND7, EPB7, EPB72
Chromosome Location
Chr.9: 121338988 - 121370304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270526.1 NP_001257455.1 2 - 3 272 121
NM_001270527.1 NP_001257456.1 2 - 3 272 121
NM_004099.5 NP_004090.4 2 - 3 272 121
NR_073037.1 3 - 4 402 121
GenBank mRNA AK302977.1 - 2 - 3 198 121
AK304449.1 - 3 - 4 346 121
AK304518.1 - 2 - 3 216 121
AK307999.1 - 2 - 3 216 121
BC010703.1 - 2 - 3 194 121
BI553227.1 - 2 - 3 201 121
BM451470.1 - - 236 121
BP208523.1 - 2 - 3 199 121
CR542100.1 - 2 - 3 155 121
DC368197.1 - 2 - 3 272 121
M81635.1 - 2 - 3 176 121
X60067.1 - 2 - 3 155 121

View Full Product Details