Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEFV
Entrez Gene ID
4210
Gene Name
Mediterranean fever
Gene Aliases
FMF, MEF, TRIM20
Chromosome Location
Chr.16: 3242028 - 3256776 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000243.2 NP_000234.1 6 - 7 1649 110
NM_001198536.1 NP_001185465.1 5 - 6 1016 110
XM_017023236.1 XP_016878725.1 6 - 7 1787 110
GenBank mRNA AF018080.1 - 6 - 7 1650 110
AK298660.1 - 6 - 7 1643 110
BC101511.1 - 6 - 7 1634 110
BC101537.1 - 6 - 7 1634 110
FJ785717.1 - 3 - 4 530 110
FJ785722.1 - 5 - 6 976 110
Y14441.1 - 6 - 7 1638 110

View Full Product Details