Product Details

Assay Reference Genome
Location

Chr.X:9739531 on build GRCh38
Cytoband
Xp22.2
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
4935
Gene Symbol
GPR143
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
Location
Chr.X:9725413-9765965 on build GRCh38
Assay Gene Location
Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GPR143 NM_000273.2 8 1222 NP_000264.2
XM_005274541.3 8 1100 XP_005274598.1
BC068977.1 8 1222 AAH68977.1
Z48804.1 8 1134 CAA88742.1

View Full Product Details