Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WBP11
Entrez Gene ID
51729
Gene Name
WW domain binding protein 11
Gene Aliases
NPWBP, PPP1R165, SIPP1, WBP-11
Chromosome Location
Chr.12: 14786478 - 14803467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016312.2 NP_057396.1 3 - 4 258 70
GenBank mRNA AB029309.1 - 3 - 4 240 70
AF118023.1 - 3 - 4 176 70
AK297410.1 - 3 - 4 282 70
AK302244.1 - 3 - 4 296 70
BC001621.1 - 3 - 4 258 70
BC023532.2 - 3 - 4 251 70
BX647347.1 - 3 - 4 267 70

View Full Product Details