Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCF2
Entrez Gene ID
4168
Gene Name
MCF.2 cell line derived transforming sequence
Gene Aliases
ARHGEF21, DBL
Chromosome Location
Chr.X: 139581768 - 139708279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099855.1 NP_001093325.1 3 - 4 300 143
NM_001171876.1 NP_001165347.1 3 - 4 300 143
XM_005262413.4 XP_005262470.1 4 - 5 448 143
XM_011531339.2 XP_011529641.1 4 - 5 507 143
XM_017029529.1 XP_016885018.1 3 - 4 269 143
XM_017029530.1 XP_016885019.1 3 - 4 269 143
XM_017029531.1 XP_016885020.1 3 - 4 202 143
GenBank mRNA AK296529.1 - 3 - 4 400 143
AK302957.1 - 3 - 4 248 143
AK316416.1 - 3 - 4 288 143
AL117234.1 - 2 - 3 183 143
DC298193.1 - 3 - 4 300 143

View Full Product Details