Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
Chromosome Location
Chr.11: 44066118 - 44084237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127219.1 NP_001120691.1 4 - 5 567 73
NM_032592.3 NP_115981.1 4 - 5 863 73
XM_005253170.3 XP_005253227.2 4 - 5 1006 73
XM_006718348.2 XP_006718411.1 4 - 5 359 73
XM_011520406.2 XP_011518708.1 4 - 5 1009 73
XM_011520407.1 XP_011518709.1 4 - 5 359 73
XM_017018421.1 XP_016873910.1 4 - 5 1010 73
GenBank mRNA AK057649.1 - 4 - 5 857 73
AK302515.1 - 4 - 5 374 73
AK307380.1 - 4 - 5 563 73
AY026508.1 - 4 - 5 818 73
BC020197.1 - 4 - 5 837 73
KU178811.1 - 3 - 4 423 73
KU178812.1 - 3 - 4 423 73
KU178813.1 - 2 - 3 363 73

View Full Product Details