Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK15
Entrez Gene ID
55554
Gene Name
kallikrein related peptidase 15
Gene Aliases
ACO, HSRNASPH
Chromosome Location
Chr.19: 50825289 - 50834337 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527087.2 XP_011525389.1 4 - 5 1203 73

View Full Product Details