The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105203.1 | NP_001098673.1 | 2 - 3 | 1589 | 71 |
NM_001105204.1 | NP_001098674.1 | 2 - 3 | 1589 | 71 | |
GenBank mRNA | AK055451.1 | - | 1 - 2 | 849 | 71 |
AK125378.1 | - | 2 - 3 | 1541 | 71 |