Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA4
Entrez Gene ID
3910
Gene Name
laminin subunit alpha 4
Gene Aliases
CMD1JJ, LAMA3, LAMA4*-1
Chromosome Location
Chr.6: 112107931 - 112254722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105206.2 NP_001098676.2 4 - 5 823 118
NM_001105207.2 NP_001098677.2 4 - 5 840 118
NM_002290.4 NP_002281.3 4 - 5 823 118
XM_005266983.3 XP_005267040.2 4 - 5 696 118
XM_005266984.3 XP_005267041.2 4 - 5 693 118
XM_017010854.1 XP_016866343.1 4 - 5 693 118
GenBank mRNA AB210027.1 - 4 - 5 675 118
AK304401.1 - 4 - 5 646 118
BC026237.1 - 4 - 5 655 118
BC066552.1 - 4 - 5 836 118
X91171.1 - 4 - 5 708 118

View Full Product Details