Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPRE3
Entrez Gene ID
22924
Gene Name
microtubule associated protein RP/EB family member 3
Gene Aliases
EB3, EBF3, EBF3-S, RP3
Chromosome Location
Chr.2: 26970371 - 27027219 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303050.1 NP_001289979.1 6 - 7 1013 87
NM_012326.3 NP_036458.2 6 - 7 1214 87
XM_006711967.3 XP_006712030.1 6 - 7 1151 87
XM_011532700.1 XP_011531002.1 6 - 7 944 87
XM_017003597.1 XP_016859086.1 6 - 7 901 87
GenBank mRNA AB025186.1 - 6 - 7 928 87
AK312342.1 - 6 - 7 948 87
BC011557.1 - 6 - 7 913 87
CR536523.1 - 5 - 6 775 87
CR541845.1 - 5 - 6 775 87

View Full Product Details