The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001752.3 | NP_001743.1 | 9 - 10 | 1283 | 95 |
GenBank mRNA | AK291585.1 | - | 9 - 10 | 1283 | 95 |
AK301577.1 | - | 8 - 9 | 1053 | 95 | |
AK315350.1 | - | 9 - 10 | 1267 | 95 | |
AY028632.1 | - | 9 - 10 | 1194 | 95 | |
BC027300.1 | - | 2 - 3 | 297 | 95 | |
BC110398.1 | - | 9 - 10 | 1272 | 95 | |
BC112217.1 | - | 9 - 10 | 1232 | 95 | |
BC112219.1 | - | 9 - 10 | 1232 | 95 | |
X04076.1 | - | 9 - 10 | 1264 | 95 |